Principaux documents
ZLXEV030WW
Milli-Q
Elix Essential 3 UV Water Purification System
About This Item
Code UNSPSC :
41104200
eCl@ss :
33050190
Nomenclature NACRES :
SB.53
Gamme de produits
Elix® Essential
Niveau de qualité
Description générale
Elix Essential systems are ideal for laboratories needing an easy-to-use economical solution that provides a constant and reliable source of Type 2 pure water. Patented Elix electrodeionization technology is combined with complementary water purification techniques, including pretreatment, reverse osmosis and bacteriocidal UV lamp treatment (in Elix Essential UV systems) to produce Type 2 pure water of consistently-pure and reliable water quality. As there are no resin packs to change, thanks to the Elix module technology, users benefit from low and predictable running costs.
Application
A pure water solution, integrating Elix technology for your essential needs
Informations légales
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Clause de non-responsabilité
Directions for Use