BMSNS1000E
Benchmark SmartDrop L Nano Spectrophotometer
AC/DC input 230 V, EuroPlug
About This Item
Code UNSPSC :
12352200
Produits recommandés
Fabricant/nom de marque
Benchmark Scientific NS1000-E
load volume
1.0-2.0 μL
Description générale
The Accuris SmartDrop series of nano spectrophotometers provides quick and easy quantification of small samples. Utilizing a small sample volume eliminates the need for dilutions, providing more accurate results. A full color, 7” touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computer is required. Each unit features a quartz pedestal onto which a drop of sample is placed. The arm is lowered and the drop is stretched into a liquid column of specificed length for spectral analysis. DNA, RNA and protein concentrations and purity numbers are displayed in a matter of seconds. The resulting data can be printed using the integrated printer, saved in the SmartDrop’s onboard memory or exported via USB as an Excel file.
Application
Spectrophotometry, OD600
Caractéristiques et avantages
- Quick and accurate results from a sample droplet
- Touch screen control panel, no PC or software required
- Built-in printer and cuvette port for OD600 readings