EP0030079345
Eppendorf® macro Vis Cuvette
max. filling volume 4,500 μL, pkg of 1000 cuvettes (10 boxes x 100 cuvettes)
About This Item
Code UNSPSC :
41121813
Nomenclature NACRES :
NB.44
Produits recommandés
Matériaux
PMMA
Caractéristiques
max. filling volume 4,500 μL
Conditionnement
pkg of 1000 cuvettes (10 boxes x 100 cuvettes)
Fabricant/nom de marque
Eppendorf® 30079345
Capacité
-4500 μL
Catégories apparentées
Description générale
“Eppendorf” macro Vis Cuvettes are single use plastic cuvettes with a maximum volume of 4500 μL.
Application
“Eppendorf” macro Vis Cuvettes are ideal for applications that go beyond the UV range, including:
- colorimetric protein assays like Bradford and Lowry methods
- determining the optical density of bacterial cultures using OD 600 methods
- as well as kinetic and fluorescence measurements.
Caractéristiques et avantages
- Macro Vis cuvettes are made of clear plastics.
- These cuvettes are ideal for measurements withing the wavelength range of 300 – 900 nm.
- “Eppendorf” macro Vis Cuvettes are specifically used in controlling temperature for kinetic measurements.
- Suitable for use with Eppendorf BioSpectrometer fluorescence and Eppendorf BioPhotometer
Informations légales
Eppendorf is a registered trademark of Eppendorf AG