Accéder au contenu
MilliporeSigma
Toutes les photos(8)

Principaux documents

ZRB2707

Sigma-Aldrich

Anti-JunB Antibody, clone 1I9-H1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonyme(s) :

Transcription factor AP-1 subunit JunB, Transcription factor JunB

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Code UNSPSC :
12352203
Nomenclature NACRES :
NA.43

Source biologique

rabbit

Niveau de qualité

Produit recombinant

expressed in HEK 293 cells

Conjugué

unconjugated

Forme d'anticorps

purified antibody

Type de produit anticorps

primary antibodies

Clone

recombinant monoclonal

Description

clone 1I9-H1
recombinant, expressed in HEK 293 cells

Gamme de produits

ZooMAb® learn more

Forme

lyophilized

Poids mol.

calculated mol wt 35.88 kDa
observed mol wt ~40 kDa

Produit purifié par

using Protein A

Espèces réactives

human, mouse

Conditionnement

antibody small pack of 25

Caractéristiques du produit alternatif plus écologique

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Validation améliorée

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

Technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

Isotype

IgG

Séquence de l'épitope

N-terminal

Numéro d'accès Protein ID

Numéro d'accès UniProt

Autre catégorie plus écologique

Conditions d'expédition

ambient

Température de stockage

2-8°C

Informations sur le gène

human ... JUNB(3726)

Description générale

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Spécificité

Clone 1I9-H1 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects JunB. It targets an epitope within 20 amino acids from the N-terminal region.

Immunogène

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human JunB.

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected JunB in A431 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected JunB in lysates from HeLa and NIH3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound JunB peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected JunB in MCF-7 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected JunB in Human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Description de la cible

Transcription factor JunB (UniProt: P17275; also known as Transcription factor AP-1 subunit JunB) is encoded by the JUNB gene (Gene ID: 3726) in human. JunB is a member of the AP-1 transcription family that binds to the DNA sequence 5′-TGA[CG]TCA-3′ and is involved in regulating gene activity following the primary growth factor response. It can bind to DNA either as a homodimer or as a heterodimer with another member of the Jun/Fos family. JunB plays an important role in proliferation, differentiation, and death. With respect to cell cycle control, JunB is reported to have a dual function. It can serve as a cell division inhibitor and a tumor suppressor or can promote cell division. It is reported to diminish cell proliferation via transcriptional activation of p16 (INK4a). Its expression is shown to be very low in quiescent cells but can be rapidly induced by mitogenic stimuli during the G0/G1 transition. It induces proliferation, survival, drug resistance in multiple myeloma (MM) cells and silencing of JunB is reported to block MM cell proliferation in a murine xenograft model. It also protects MM cells against dexamethosome and bortezomib-induced cell death. JunB is also reported to regulate epidermal stem cells and sebaceous glands through balancing proliferation and differentiation of progenitors and by suppressing lineage infidelity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Singh, K., et al. (2018). Nat. Commun. 9; Article 3425; Fan, F., et al (2017). Leukemia. 31(7); 1570-1581; Farras, R., et al. (2008). Mol. Cell. Biol. 28(12); 4173-4187; Passegue, E., and Wagner, EF. (2000). EMBO J. 19(12); 2969-2979).

Forme physique

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Stockage et stabilité

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Informations légales

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Clause de non-responsabilité

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Vous ne trouvez pas le bon produit ?  

Essayez notre Outil de sélection de produits.

Code de la classe de stockage

11 - Combustible Solids

Classe de danger pour l'eau (WGK)

WGK 1

Point d'éclair (°F)

Not applicable

Point d'éclair (°C)

Not applicable


Faites votre choix parmi les versions les plus récentes :

Certificats d'analyse (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

Si vous avez besoin d'assistance, veuillez contacter Service Clients

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique