940313
Vulcan® XC72

conductive carbon black
Synonym(s):
Black pearl, Vulcan XC-72
grade
battery grade
Quality Level
form
powder
pH
~250(m2/g (BET))
bp
4200 °C
mp
3550 °C
bulk density
~0.265 g/cm3
application(s)
battery manufacturing
battery manufacturing
SMILES string
[C]
General description
Vulcan XC72 is a mesoporous carbon black characterized by its moderately high surface area, approximately 250 m²/g, coupled with superior electrical conductivity. The carbon particles exhibit an approximately spherical morphology and possess a primary size distribution ranging from 30 to 60 nanometers. This unique combination of features makes Vulcan XC72 an ideal candidate for various applications requiring antistatic and conductive properties.
Application
VULCAN XC72 is widely used in research and development for numerous antistatic and conductive purposes, particularly as a catalyst support. Its high specific surface area and excellent electrical conductivity contribute to enhanced performance in proton-exchange membrane fuel cells (PEMFC), outperforming materials such as acetylene black. Vulcan XC72′s adaptability facilitates the creation of catalyst composites, where catalyst nanoparticles may be either uniformly dispersed and deposited onto the carbon black substrate or synthesized in situ. The material′s extensive surface area is instrumental in regulating the size of catalyst nanoparticles, with evidence suggesting that carbon blacks with larger surface areas tend to produce smaller nanoparticles. Beyond fuel cell applications, Vulcan XC72 has also been employed as a catalyst support in various battery technologies, including redox flow batteries and metal-air batteries, such as zinc-air and sodium-air, where its properties enhance the overall electrochemical performance.
Legal Information
Vulcan is a registered trademark of Cabot Corporation
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.