Skip to Content
Merck

1.46797

Tryptic Soy Agar - Ready-to-use Contact Plates

Tween®, Sodium thiosulfate, Histidine, Lecithin, ICR plate with non-lockable lid, plate diam. 55 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for surface monitoring

Synonym(s):

CASO agar, Casein Soybean Digest Agar with neutralizer, TSA, Tryptic Soy Agar, Tryptic Soy Agar with Lecithin, Tween, Histidine and Thiosulfate, Tryptone soy agar with neutralizers

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
41106231
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

polystyrene plate

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate (ready to use culture media)

feature

2D bar code
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

packaging

pkg of 10 plates Triple packed

plate diam.

55 mm

application(s)

microbiology
pharmaceutical

surface monitoring

storage temp.

2-25°C

suitability

nonselective for

General description

Tryptic Soy Agar is a complex nutrient rich medium designed to determine the total microbial count ( bacteria, yeast and molds) on dry and sanitized surfaces of isolators and clean rooms. The medium is supplemented with pyruvate, which efficiently neutralizes the presence of hydrogen peroxide used during decontamination of isolators. The neutralizers lecithin, polysorbate Tween®80, histidine, and sodium thiosulfate inactivate possible active agents of disinfectants. The plates are triple bagged, and gamma irradiated in the final packaging and used for surface and personnel monitoring in sterile production area.

Application

Tryptic Soy Agar with LTHThio - ICR contact plates are designed to monitor and determine the total microbial count on dry, sanitized surfaces and personnel in isolators and clean rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service