FLUH99C76F9C
Fluorochem (Preferred Partner)
5-(Methylsulfonyl)-indoline
Iupac Name
5-(methylsulfonyl)indoline
Inchi Code
1S/C9H11NO2S/c1-13(11,12)8-2-3-9-7(6-8)4-5-10-9/h2-3,6,10H,4-5H2,1H3
InChI key
OFYOHQMIBTVTKY-UHFFFAOYSA-N
storage temp.
Ambient Storage
Purity
95.00%
Country of Origin
GB
Physical Form
Solid
Product Link
https://shop.fluorochem.co.uk/p/F023743
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/sds/sds_F023743_en.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.