Skip to Content
Merck

ZRB2989

Anti-PENK Antibody, clone 1M12 ZooMAb® Rabbit Monoclonal

newenhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

Enkephalin

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1M12, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30.79 kDa
observed mol wt ~32 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PENK(5179)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1M12 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects Proenkephalin-A (PENK). It targets an epitope within 24 amino acids from the second PENK peptide in the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the second PENK peptide of human Proenkephalin-A.

Application

Quality Control Testing

Evaluated by Immunohistochemistry in human adrenal gland tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Proenkephalin-A (PENK) in human adrenal gland tissue sections.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PENK in SH-SY5Y cells.

Western Blotting Analysis: A 1:50,000 dilution from a representative lot detected His-tagged, full length recombinant of human Proenkephalin-A (PENK).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Proenkephalin-A (UniProt: P01210) is encoded by the PENK gene (Gene ID: 5179) in human. Proenkephalin-A is a precursor protein that gives rise to enkephalins, which are endogenous opioid peptides involved in pain modulation and various physiological processes. It consists of a long polypeptide chain that is post-translationally processed at 8 cleavage sites to produce multiple enkephalin peptides (Syn-enkephalin, Met-enkephalin, Leu-enkephalin) and three PENK peptides (aa 114-133, aa 143-183, aa 237-258). These peptides take part in controlling learning and memory, analgesia and other brain-regulated behaviors. Proenkephalin-A is predominantly expressed in the central nervous system, particularly in regions such as the brain and spinal cord. It is also found in the adrenal medulla and various peripheral tissues, indicating its role in both central and peripheral pain pathways. It is primarily localized in the cytoplasm of neurons, where it is synthesized and processed. After translation, it is transported to secretory vesicles, where it undergoes further processing to produce active enkephalin peptides. Alterations in Proenkephalin-A expression or function have been implicated in various diseases, including chronic pain conditions, addiction, depression, and anxiety disorders. The gene is shown to be hyper-methylated in prostate cancer and thus may be of use as a molecular marker for prostate cancer diagnosis. Increased plasma PENK concentrations are found to be associated with long-term kidney outcomes and mortality. Studies report regulatory role of PENK-A as a brake in kidney regeneration. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Khorashadi, M., et al. (2020). Nephron. 144(12):655-661; Chi, Liu., et al. (2023). Nature Communications. 14:7167).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service