Skip to Content
Merck

MABT1576

Anti-Siglec-15 Antibody, clone MK4.5

new

Synonym(s):

SIGLEC-I, Sialic acid binding Ig-like lectin 15

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MK4.5, monoclonal

mol wt

calculated mol wt 36.92 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human, mouse

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

mouse ... Siglec15(620235)

Specificity

Clone MK4.5 is a mouse monoclonal antibody that specifically detects Siglec-15. It targets an epitope within an Extracellular Ig-like 1-2 domain.

Immunogen

Murine Siglec-15 Fc fusion protein.

Application

Quality Control Testing

Evaluated by Flow Cytometry in RAW 264.7 cells.

Flow Cytometry Analysis: 1.0 µg from a representative lot detected Siglec-15 in one million RAW 264.7 cells treated with 180 ng/mL of human TNF-related activation-induced cytokine (TRANCE) for 4 days.

Tested Applications

Immunofluorescence Analysis: A representative lot detected Siglec-15 in Immunofluorescence application (Korn, M.A., et al. (2020). J Immunol. 205(10):2595-2605).

Flow Cytometry Analysis: A representative lot detected Siglec-15 in Flow Cytometry application (Korn, M.A., et al. (2020). J Immunol. 205(10):2595-2605).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Siglec-15 (UniProt: A7E1W8; also known as SIGLEC-I, Sialic acid binding Ig-like lectin 15) is encoded by the Siglec15 gene (Gene ID: 620235) in mouse. SIGLEC15 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 24-342), a transmembrane domain (aa 263-283). In human Siglec-15 (UniProt: Q6ZMC9; also known CD33 antigen-like 3) expression is observed in macrophages and dendritic cells of spleen and lymph nodes. Siglec-15 serves as an immune checkpoint molecule and a critical immune suppressor. It suppresses antigen-specific T cell responses in vitro and in vivo. Genetic ablation or antibody blockade of SIGLEC15 is reported to amplify anti-tumor immunity in the tumor microenvironment and inhibits tumor growth in murine models. SIGLEC15-deficient mice also exhibit decreased serum IL-10 levels. Siglec-15 is suggested to be a potential regulator of osteoporosis, cancer, and some infectious diseases. The study with this MK4.5 mouse monoclonal antibody demonstrated an almost exclusive Siglec-15 expression on murine osteoclasts and hardly any other expression on various other immune cell types whereas, human Siglec-15 was more broadly expressed on human myeloid cells, including human osteoclasts. Findings suggest a role of Siglec-15 as a regulator of pathologic bone resorption in arthritis and highlight its potential as a target for future therapies. This MK4.5 antibody has been validated in Flow cytometry and Immunofluorescence. (Ref.: Korn, M.A., et al. (2020). J Immunol. 205(10):2595-2605).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service