Skip to Content
Merck
All Photos(1)

Key Documents

591874

Sigma-Aldrich

Poly(2,2,3,4,4,4,-hexafluorobutyl methacrylate-co-glycidyl methacrylate)

glycidyl methacrylate 50 mol %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H8F6O2)M(C7H10O3)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

composition

glycidyl methacrylate, 50 mol %

solubility

DMSO: soluble
NMP: soluble
THF: soluble
methylene chloride: soluble

Looking for similar products? Visit Product Comparison Guide

General description

Transparent, low surface tension polymer.

Application

Low refractive index, photocrosslinkable polymer for cladding layer in optical waveguides.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service