Skip to Content
Merck
All Photos(1)

Key Documents

939668

Sigma-Aldrich

TissueFab® norbornene functionalized poly(amino acid)

new

hydrogel and bioink precursor, for tissue engineering

Synonym(s):

Amino acid, Amino acid polymer, Aspartic acid, Bioink precursor polymers, Hydrogel materials, Norbornene functionalized polymer, Poly(aspartic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H14N2O4)m(C12H16N2O2)n
UNSPSC Code:
12352200

Quality Level

form

powder or chunks

mol wt

Mw 50-120 kDa by GPC

color

white

application(s)

(Tissue engineering, 3D bioprinting, 3D cell culture)

compatibility

cell culture

storage temp.

−20°C

General description

Rethink your bioinks with TissueFab®

About the material:

These TissueFab® norbornene functionalized poly(amio acid) is a xeno-free (animal origin free), biodegradable and biocompatible polymer that been developed and tested for hydrogel synthesis for tissue engineering and other biomedical applications. Printing and crosslinking parameters:

  • Compatible for cell encapsulation and hydrogel formation.
  • Crosslinked via photopolymerization via UV or visible light with suitable photo initiator.
The TissueFab® norbornene functionalized poly(amino acid) is characterized by its biocompatibility and biodegradability, making it well-suited for a broad spectrum of applications in the biomedical field. This poly(amino acid) is synthesized through the ring opening of polysuccinimide using a nucleophilic reagent. The presence of amide linkages in these polymers results in peptide-like structures that are prone to cleavage and degradation in the body, leading to the formation of amino acids or small peptides.

Furthermore, the norbornene functional group within the polymer provides sites for additional chemical modifications or conjugation with bioactive molecules, such as therapeutics, peptides, and targeted ligands. This versatility enables the incorporation of specific functionalities into the polymer matrix, allowing for the impartation of desired biological or therapeutic properties, such as controlled drug release or cell adhesion. For instance, the norbornene polymer can undergo thiolene reactions with thiolated peptides, thereby eliciting bioactive biochemical cues to encapsulated cells.

More information and a step-by-step protocol can be found under the "More Documents" section of this page.

Application

The TissueFab® norbornene functionalized poly(amino acid) xeno-free polymer is:
  • Biocompatible and biodegradable precursor for hydrogel and bioink development
  • Suitable for a wide range of biomedical applications, including 3D cell culture, tissue engineering, in vitro drug screening, and regenerative medicine research
  • Enables the development of xeno-free polymers for applications in advanced cell culture and tissue engineering
  • Ideal for creating biocompatible materials that support cell growth and tissue regeneration in a xenogeneic-free environment

Features and Benefits

This TissueFab® hydrogel and bioink precursor has been developed and tested to meet strict criteria to best suit researchers′ needs. This includes:

  • Xeno-free, animal-origin free
  • Tunable
  • Cell viability
  • Biocompatibility and biodegradability
  • Mechanical and rheological properties
  • Crosslinkability

Legal Information

TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service