Skip to Content
Merck
All Photos(1)

Key Documents

939854

Sigma-Aldrich

Gelatin norbornene

new

Synonym(s):

Norbornene functionalized gelatin, Norbornene gelatin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C36H52N9O11)n
UNSPSC Code:
12352200

form

powder or chunks (or fibers)

Quality Level

color

white to light yellow-brown

storage temp.

2-8°C

SMILES string

CC(C(NCC(N1CCCC1C(NC(CCCCNOC(C2C(C3)C=CC3C2)=O)C(NCC(NC(CCC([O-])=O)C(N4C(C(N[*])=O)CC(O)C4)=O)=O)=O)=O)=O)=O)N[*]

General description

Our gelatin norbornene is a functionalized natural polymer that combines the biocompatibility and biodegradability of gelatin with the click-chemistry capabilities of norbornene. By leveraging the unique properties of gelatin and modifying it with norbornene functional groups, we have developed a versatile biomaterial that offers tailored solutions for various biomedical applications.

Gelatin norbornene, characterized by primary amines serving as nucleophiles for reaction with carbic anhydride to form amide-linked norbornene, holds great promise for tissue engineering, regeneration, and stem cell culture. The highly reactive nature of norbornene groups with thiols enables light-mediated orthogonal or step growth reactions, leading to the formation of thiol-norbornene (thiol-ene) crosslinks. The resulting hydrogels exhibit faster gelation and superior cytocompatibility, making them particularly favorable for cell-based applications.

Application

With its ability to mimic the naturally occurring extracellular matrix of tissues and the capability to create unique and tailorable gelatin scaffolds and hydrogels, gelatin norbornene stands as a game-changing solution for advancing biomedical research. Applications include:
  • Studying cell motility in 3D cell culture scaffolds
  • Engineering biocompatible scaffolds and hydrogels for areas such as cartilage tissue engineering, bone tissue engineering, and neural regeneration
  • Creating in vitro models for studying disease, development or for use in drug screening applications
  • Therapeutic drug or cell encapsulation and delivery
  • Therapuetic conjugation
The nobornene funtionalized gelatin can also be used to create complex functionalized materials through the tethering of cysteine or thiol containing biomolecules such as peptides, growth factors, and other proteins.

Features and Benefits

  • Biocompatible and biodegradable.
  • Fast gelation through light mediated click chemistry.
  • 30-50% functionalized.
  • Tailorable for tethering additional functional groups or biochemical moieties or adjusting 3D hydrogel biochemical or mechanical properites.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service