Skip to Content
Merck
All Photos(1)

Key Documents

BR937241

BRAND® BLAUBRAND® volumetric flasks, PP stopper, ISO

new

class A, volume 10 mL, flask joint: ST/NS 7/16, Boro3.3

Synonym(s):

BRAND BLAUBRAND, analytical flask, calibration flask, class A glassware, glassware, volumetric flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804

material

borosilicate glass 3.3 flask
polypropylene stopper

Joint

joint: ST/NS flask7/16

feature

certificate ISO
class A
closure type stopper

packaging

pack of 1 ea

manufacturer/tradename

BRAND 937241

volume

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

Volumetric flasks, class A/AS/AW, are calibrated to the highest-grade accuracy and are used for the preparation of highly accurate dilutions and standard solutions.

Application

Liquid handling, dilutions, titrations, chemical analysis, reagent preparation

Features and Benefits

  • Class AS
  • DE-M marking Boro 3.3
  • DIN EN ISO 1042. Calibrated to contain (TC, In)
  • Includes ISO individual certificate
  • Polypropylene stopper

Legal Information

BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service