BR937243
BRAND® BLAUBRAND® volumetric flasks, PP stopper, ISO

class A, volume 10 mL, flask joint: ST/NS 10/19, Boro3.3
Synonym(s):
BRAND BLAUBRAND, analytical flask, calibration flask, class A glassware, glassware, volumetric flask
About This Item
Recommended Products
material
borosilicate glass 3.3 flask
polypropylene stopper
Joint
joint: ST/NS flask10/19
feature
certificate ISO
class A
closure type stopper
packaging
pack of 1 ea
manufacturer/tradename
BRAND 937243
volume
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Class AS
- DE-M marking Boro 3.3
- DIN EN ISO 1042. Calibrated to contain (TC, In)
- Includes ISO individual certificate
- Polypropylene stopper
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service