Skip to Content
Merck
All Photos(1)

Key Documents

700381J

Avanti

Geranylgeranoic acid

new

Avanti Research - A Croda Brand

Synonym(s):

(2E,6E,10E)-3,7,11,15-tetramethyl-2,6,10,14-hexadecatetraenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O2
Molecular Weight:
304.47
MDL number:
UNSPSC Code:
12352200

biological source

synthetic

Assay

>99% (TLC)

form

methyl acetate solution

packaging

pkg of 1 × 10 mg (700381J-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(O)C=C(C)CCC=C(C)CCC=C(C)CCC=C(C)C

InChI key

SZNLKILVMCHHSD-UHFFFAOYSA-N

Application

lipids research

Packaging

2 mL Amber Glass Sealed Ampule (700381J-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service