197224
Bcr-abl Inhibitor III, GNF-5
The Bcr-abl Inhibitor III, GNF-5 controls the biological activity of Bcr-abl. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Bcr-abl Inhibitor III, GNF-5, Abl Inhibitor III, N-(2-Hydroxyethyl)-3-(6-(4-(trifluoromethoxy)phenylamino)pyrimidin-4-yl)benzamide, N-(2-Hydroxyethyl)-3-(6-(4-(trifluoromethoxy)phenylamino)pyrimidin-4-yl)benzamide, Abl Inhibitor III
Select a Size
₹17,369.00
Select a Size
About This Item
₹17,369.00
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 50 mg/mL, clear, colorless
shipped in
ambient
storage temp.
2-8°C
1 of 4
This Item | 676504 | 361555 | 648451 |
---|---|---|---|
form powder | form powder | form solid | form powder |
assay ≥97% (HPLC) | assay ≥97% (HPLC) | assay ≥95% (HPLC) | assay ≥98% (HPLC) |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
storage condition OK to freeze, protect from light | storage condition OK to freeze, protect from light | storage condition OK to freeze, protect from light | storage condition OK to freeze, protect from light |
General description
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service