Skip to Content
Merck
All Photos(1)

Key Documents

5.04187

Sigma-Aldrich

P2Y Antagonist II, BPTU

Synonym(s):

P2Y Antagonist II, BPTU, Purinergic Receptor P2Y Antagonist II, 1-(2-(2- tert-Butylphenoxy)pyridin-3-yl)-3-(4-(trifluoromethoxy)phenyl)urea, Purinergic Receptor P2Y Antagonist II, 1-(2-(2-tert-Butylphenoxy)pyridin-3-yl)-3-(4-(trifluoromethoxy)phenyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22F3N3O3
CAS Number:
Molecular Weight:
445.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1ccc(cc1)NC(=O)Nc2c(nccc2)Oc3c(cccc3)C(C)(C)C

InChI

1S/C23H22F3N3O3/c1-22(2,3)17-7-4-5-9-19(17)31-20-18(8-6-14-27-20)29-21(30)28-15-10-12-16(13-11-15)32-23(24,25)26/h4-14H,1-3H3,(H2,28,29,30)

InChI key

AHFLGPTXSIRAQK-UHFFFAOYSA-N

General description

A phenoxypyridinyl-phenylurea that acts as a highly selective P2Y1 antagonist (Ki = 6 nM against 0.5 nM [β-33P]-2-MeS-ADP in competitive binding assays toward human P2Y1), while displaying much reduced or no affinity toward other P2Y family G-protein-coupled purinergic receptors (Ki = ≥2.5 µM against 0.5 nM [β-33P]-2-MeS-ADP binding toward hP2Y14; Ki >15 µM toward human P2Y2, P2Y6, P2Y11, and P2Y12). Effectively inhibits 2.5 µM ADP-induced platelet aggregation in vitro (IC50 = 2.1 µM using human platelet rich plasma) and reduces FeCl2-induced (via 10 min localized artery surface exposure to 50% FeCl2) carotid artery internal thrombus (blood clot) formation in anesthetized rats in vivo (by 68% 50 min post 10 min FeCl2 exposure; single 10 mg/kg i.v. 15 min prior to FeCl2 exposure, followed by continuous i.v. infusion at 10 mg/kg/h) with only 3.3- and 3.1-fold enhanced cuticle and mesenteric bleeding time, respectively. The same antithrombosis efficacy when achieved with the P2Y12 antagonist Clopidogrel (20 mg/kg p.o.) is reported to result in much prolonged cuticle and mesenteric bleeding time (4.1- and 8.2-fold of control level, respectively). Pharmacokinetic studies in rats indicate a moderate oral bioavailability (F = 10%; Cmax = 5.83 µM; Tmax = 2 h; T1/2 = 1.43 h; 30 mg/kg delivered with 10% cremophor:10% EtOH:80% H2O).

Biochem/physiol Actions

Primary Target
P2Y₁
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhang, D., et al. 2015. Nature In press.
Chao, H., et al. 2013. J. Med. Chem.56, 1704.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service