Skip to Content
Merck
All Photos(1)

Key Documents

5.08942

Sigma-Aldrich

Prokineticin 2 Antagonist, PKRA7

Synonym(s):

Prokineticin 2 Antagonist, PKRA7, PROK2 Antagonist, PKRA7, Bv8 Antagonist, PKRA7, PK2 Antagonist, PKRA7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34ClFN2O4
CAS Number:
Molecular Weight:
505.02
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

semisolid

potency

5 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell permeable, brain permeant 3-pyrrolidine carboxamide derivative that acts as an antagonist of polypeptide chemokine prokineticin 2 (PK2) receptor (Bv8, PROK2; IC50 = 5 nM and 8.2 nM for PKR1 and PKR2, respectively). Suppresses the growth of D456MG glioma cell xenografts in nude mice by reducing blood vessel density and blocking angiogenesis via inhibition of PKR1 and PKR2 expressed on endothelial cells. Shown to suppress the growth of AsPC1 and CFPac-1 pancreatic tumor cells in xenograft model of nude mice by reducing macrophage migration and inflitration. Reduces the expression of CCL27, CCR10, CCR4, CCR5, and CCR6 pro-migratory chemokines and chemokine receptors in macrophages. Enhances the efficacy of standard temomolomide in the treatment of glioblastoma and gemcitabine in the treatment of pancreatic cancer in xenograft nude mice models.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
PROK2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Curtis, V., et al. 2013. PLoS One.8, e54916.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service