Skip to Content
Merck
All Photos(1)

Key Documents

5.34339

Sigma-Aldrich

PTPσ Inhibitor, ISP

Synonym(s):

PTPσ Inhibitor, ISP, Protein Tyrosine Phosphatase σ Inhibitor, ISP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C177H306N66O54S3
Molecular Weight:
4318.93
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 100 mg/mL
water: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable, TAT conjugated peptide derived from the protein tyrosine phosphatase sigma (PTPsigma) wedge domain that binds to PTPsigma and relieves glial-derived chondroitin sulfate proteoglycan (CSPG)-mediated inhibition. Shown to cross the blood-brain barrier. Does not bind to other CSPG receptors, such as LAR and NgRs. By blocking the PTPsigma-CSPG interaction it abolishes CSPG -mediated inhibition of axonal regrowth and sprouting. Shown to promote adult sensory neuron growth cone (~ 2.5 µM). Restores serotonergic innervations to the spinal cord below the level of injury and promotes progressive functional recovery of both locomotor and urinary systems after spinal cord injury in a rat model (11 µg/day).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PTPσ
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

NH2-GRKKRRQRRRCDMAEHMERLKANDSLKLSQEYESI-NH2

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lang, B. T, et al. 2015. Nature.518, 404.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service