Skip to Content
Merck
All Photos(1)

Key Documents

6.18283

Sigma-Aldrich

1,4-Dichlorobenzene

for synthesis

Synonym(s):

1,4-Dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
1,4-(Cl)2C6H4
CAS Number:
Molecular Weight:
147.00
MDL number:
UNSPSC Code:
12352100

grade

for synthesis
synthesis grade

Quality Level

SMILES string

Clc1ccc(cc1)Cl

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

OCJBOOLMMGQPQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service