Skip to Content
Merck
All Photos(1)

Key Documents

6.18400

Supelco

Benedict′s reagent

qualitative

Synonym(s):

Benedict’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302

SMILES string

[Cu+2].[Na+].[Na+].[S](=O)(=O)(O)O.[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].[O-]C(=O)O.O

InChI

1S/C6H8O7.CH2O3.Cu.2Na.H2O4S.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;2-1(3)4;;;;1-5(2,3)4;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);(H2,2,3,4);;;;(H2,1,2,3,4);1H2/q;;+2;2*+1;;/p-4

InChI key

QFKWSRIUZIYLCK-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service