Skip to Content
Merck
All Photos(1)

Key Documents

6.18553

Supelco

Potassium dihydrogen orthophosphate

for chromatography

Synonym(s):

Dihydrogen potassium phosphate, Monobasic potassium phosphate, Sorensen′s potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161706

grade

for chromatography

suitability

suitable for chromatography

Application


  • Fluoride contents in commonly used commercial phosphate fertilizers: This research uses potassium dihydrogen orthophosphate to evaluate the fluoride content in phosphate fertilizers, addressing environmental concerns associated with fluoride pollution. The findings are essential for environmental monitoring and ensuring agricultural product safety (Li H et al., 2023).

  • The role of phosphorus speciation of biochar: Investigating the effects of potassium dihydrogen orthophosphate on the speciation of phosphorus in biochar, this study explores its potential to reduce heavy metal availability in soils. Such applications are critical for remediating polluted environments and enhancing soil health (Huang K et al., 2023).

  • Observation of the phase-separation multiphase flow: Utilizing potassium dihydrogen orthophosphate in studying phase separation in multiphase flow systems, this research contributes significantly to material science, particularly in the analysis of complex fluid dynamics, which is vital for developing advanced materials and processing techniques (Nishimura K et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service