Skip to Content
Merck
All Photos(1)

Key Documents

6.18558

Supelco

Di-Sodium hydrogen Phosphate

for chromatography, LiChropur

Synonym(s):

Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12352302
grade:
for chromatography

grade

for chromatography

product line

LiChropur

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

suitability

suitable for chromatography

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service