Skip to Content
Merck

6.18615

Sigma-Aldrich

tert-Butyl Methyl Ether

for chromatography

Synonym(s):

tert-Butyl methyl ether

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12352112
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

for chromatography

Quality Level

suitability

suitable for chromatography

SMILES string

O(C(C)(C)C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butylmethyl ether, also known as MTBE, is added to gasoline to increase its oxygen content. It is used as a solvent in gas chromatography (GC) for the analysis of various compounds, including gasoline components and their degradation products.

Application


  • Bromate ion chromatography standard: Employed in the trace determination of disinfection by-products in drinking water, enhancing water safety assessments and regulatory compliance using cyclic ion chromatography with large-volume direct injection (Zhu et al., 2023).

  • Pharmaceutical bromate analysis: Used in the ultra-trace determination of oxyhalides in marine waters treated with ozone, vital for environmental monitoring and ensuring aquacultural water quality using direct injection ion chromatography coupled with mass spectrometry (Sanz Rodriguez et al., 2021).

  • IC bromate calibration standard: Applied in the safety evaluation of bottled water, specifically for the determination of bromate and other oxyhalides, to meet health standards and improve public safety measures (Djam et al., 2020).

  • Bromate detection in biotech: Utilized in direct injection methods for robust analysis of trace-level bromate and bromide in drinking water, crucial for ensuring compliance with health regulations (Lawal et al., 2010).

  • High-purity bromate IC reagent: Integral in the rapid simultaneous analysis of iodoacetic acids, bromoacetic acids, bromate, and other halogenated compounds in water, supporting environmental monitoring and toxicological research (Shi et al., 2009).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service