Skip to Content
Merck
All Photos(1)

Key Documents

6.18717

Supelco

Silicone Oil

Synonym(s):

Silicone oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23201100

Application


  • An eco-friendly and facile method for oil-water separation using the bio-Zn oxide-based superhydrophobic membrane.: This research highlights the application of silicone oil in developing superhydrophobic membranes for efficient oil-water separation. The study focuses on the environmental benefits and the ease of application, demonstrating the role of silicone oil in enhancing the hydrophobicity and efficiency of bio-Zn oxide-based membranes. This aligns with current trends in sustainable technology and water purification methods (Beagan et al., 2024).

  • Tuning contact line dynamics on slippery silicone oil grafted surfaces for sessile droplet evaporation.: This paper discusses the manipulation of silicone oil properties to modify surface interactions in laboratory settings, specifically for controlling the evaporation dynamics of droplets. The research contributes to a deeper understanding of fluid dynamics on coated surfaces, which has significant implications for analytical methodologies in chemistry and materials science (Raynard et al., 2024).

  • Super-Low Friction Electrification Achieved on Polytetrafluoroethylene Films-Based Triboelectric Nanogenerators Lubricated by Graphene-Doped Silicone Oil.: This study leverages silicone oil enhanced with graphene to reduce friction in triboelectric nanogenerators, showcasing its utility in enhancing the performance and durability of energy harvesting devices. Such applications underscore the role of silicone oil in emerging technologies and its potential for broader industrial and environmental applications (Chen et al., 2023).

  • Self-Healing, Robust, Liquid-Repellent Coatings Exploiting the Donor-Acceptor Self-Assembly.: The research explores the use of silicone oil in creating advanced materials with self-healing and liquid-repellent properties. This application highlights the intersection of silicone oil with materials science to develop coatings that could revolutionize surface technologies and materials engineering (Zhang et al., 2023).

  • Surfactant-laden droplet size prediction in a flow-focusing microchannel: a data-driven approach.: This paper presents a novel application of silicone oil in microfluidic systems to control droplet formation and stability. The study integrates a data-driven approach to predict and manipulate droplet sizes, which is crucial for applications in chemical synthesis, drug delivery systems, and diagnostic technologies (Chagot et al., 2022).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

599.0 °F

Flash Point(C)

315 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service