Skip to Content
Merck
All Photos(1)

Key Documents

71241-M

Sigma-Aldrich

pIEx-1 DNA - Novagen

Novagen′s pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodopteraderived insect cells.

Synonym(s):

InsectDirect System- pIEx-1 DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

dry ice
wet ice

storage temp.

−20°C

General description

The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. pIEx-1 contains N-terminal His•Tag and S•Tag coding sequences, and a C-terminal HSV•Tag coding sequence. N-terminal vector encoded sequences can be completely removed by cloning into the PshA I site and cleaving the fusion protein with enterokinase.

pIEx-1 is also available as an Ek/LIC vector. The Ek/LIC vector is prepared for rapid directional cloning of PCR-amplified DNA. Using specifically designed primers for amplification, inserts can be efficiently cloned without the need for restriction enzyme digestion or ligation.

For sequencing or PCR of recombinants in pIEx-1, refer to the table below to choose the appropriate primers.

Warning

Toxicity: Standard Handling

Other Notes

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.

Legal Information

INSECTDIRECT is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service