Skip to Content
Merck
All Photos(1)

Key Documents

Y0001849

Erythromycin stearate for impurity S identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Erythromycin stearate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H67NO13 · C18H36O2
CAS Number:
Molecular Weight:
1018.40
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

erythromycin

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O.CC[C@@H]1OC(=O)[C@@H](C)[C@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@@H](C)[C@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(O)C[C@H](C)C(=O)C(C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H67NO13.C18H36O2/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;2-17H2,1H3,(H,19,20)/t18-,19+,20?,21+,22-,23-,24-,25-,26-,28+,29+,30+,31-,32-,34-,35-,36+,37?;/m0./s1

InChI key

YAVZHCFFUATPRK-JGJPHTNUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 15 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service