Skip to Content
Merck
All Photos(1)

Key Documents

08081401

Sigma-Aldrich

MT-2

from human lymph

Synonym(s):

MT 2, MT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.81

biological source

human lymph

form

liquid

growth mode

Non-adherent or adherent on poly-L-lysine treated plastic

karyotype

2n=46

morphology

Lymphoblast

products

Adult T-cell leukaemia virus (ALT; HTLV-1)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

-65 to -96°C

Cell Line Origin

Human T cell leukaemia

Cell Line Description

Derived from a 45 year old Japanese female with generalised lymphadenopathy and heptosplenomegaly (Adult T-cell Leukaemia) by co-culture of her peripheral leukocytes with male umbilical cord lymphocytes. The cells carry HTLV-1 and support the growth of HIV. The cells should be handled under laboratory containment level 3 conditions. MT-2 has a male karyotype and the presence of a Y chromosome has been confirmed by STR profiling at ECACC

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% Decomplemented Fetal Bovine Serum (FBS).

Subculture Routine

Seed cultures between 2-3x100,000 cells/ml. Saturation density 9 x 100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service