Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR02CPO

PCR strip tube caps

thin wall, 8 per strip, size 0.2 mL, orange, pkg of 10x125strips/cs

Synonym(s):

Corning tube cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

manufacturer/tradename

Axygen Inc PCR-02CP-O
Corning

packaging

pkg of 10x125strips/cs

size

0.2 mL

color

orange

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service