AXYTE1004BL
MAXYMum Recovery® pipette tips for Eppendorf® and Socorex Pipettors
size 1000 μL, blue, pkg of 10x500tips/cs
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1000 μL
packaging:
pkg of 10x500tips/cs
manufacturer/tradename:
Axygen Inc TE-1004-B-L
Corning
Corning
manufacturer/tradename
Axygen Inc TE-1004-B-L
Corning
packaging
pkg of 10x500tips/cs
size
1000 μL
volume
1000 μL
color
blue
General description
Products are available in China only.
Other Notes
Browse all pipette tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service