Skip to Content
Merck
All Photos(2)

Key Documents

BMSMR9610E

Benchmark SmartReader UV-Vis Microplate Absorbance Reader

new

AC/DC input 230 V, EuroPlug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

manufacturer/tradename

Benchmark Scientific MR9610-E

parameter

at ambient + 5 -45 °C
200-1000 nm (Wavelength range)

W × D × H

-300 mm -11.8 in. -500 mm -19.7 in. -260 mm -10.25 in.

Looking for similar products? Visit Product Comparison Guide

General description

The Accuris SmartReader UV-Vis Microplate Readers (models MR9610 and MR9611) have been designed with ease-of-use and fast results in mind. These instruments brings extensive capabilities to the lab bench and are designed for the busy lab with multiple users. A large touch screen command center is extremely user-friendly and allows for fast set up of new protocols, as well as modifying and running existing protocols.The monochromator-based optical system allows for precise selection of wavelengths from 200nm to 1000nm, with no filters required. This range makes the SmartReader UV-Vis useful not only for typical ELISA , Bradford Lowry, and turbidity assays but also for more advanced applications such as quantitation of nucleic acids and proteins. The SmartReader UV-Vis can be set for Endpoint, Kinetic and Spectral Scanning assays.The MR9610 is designed for efficient processing of multiple samples and accepts standard 96 well plates as well as 384 well plates for high density screening. The plate layout interface allows for a quick and intuitive setup of a plate well’s contents. Users can highlight and label multiple wells to save time. The display shows a graphical representation of the plate. Users can label each well as required for: “known standard”, “unknown sample”, “blank well” , “QC Control well”, “positive control”, “negative control”, and “empty”.Controllable plate shaking and incubation allow for optimization of assay results. The shaking speed can be controlled at different intensities with options for timed and pulsed shaking before a reading, or in between readings when performing a kinetic assay. The linear motion efficiently and uniformly mixes all samples in both 96 and 384 well plates. Temperature can be controlled from ambient +5⁰C up to 45⁰C, allowing for the processing of enzyme kinetics, and bacterial growth assays.

Application

ELISA, Absorbance readings, Spectral Scanning, End Point, Kinetic, Microvolume Nucleic Acid/Purity Measurements, etc.

Features and Benefits

  • 10.0 inch color touch screen for programming & operation
  • Accepts 96 & 384 well plates with/without lids
  • MR9610-SDP allows for low microvolume measurements for determining Nucleic Acid / Protein concentration & purity

Legal Information

SmartReader is a trademark of Benchmark Scientific, Inc.

accessory


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service