DWK218013651
Duran® laboratory bottles, with caps

clear, graduated, neck joint: threaded GL45, pouring ring, bottle capacity 250 mL
Synonym(s):
218013651, DURAN lab bottle, Duran® laboratory bottles, with caps, lab bottle, laboratory bottle, media bottle, storage bottle
material
blue polypropylene cap
borosilicate glass 3.3 bottle
clear
Joint
joint: threaded neck GL45
sterility
non-sterile
feature
autoclavable
cap
graduated
pouring ring
packaging
pack of 10 ea
manufacturer/tradename
DWK Life Sciences DURAN® 218013651
bottle capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Complies with the standards in ISO 4796-1:2016 Laboratory glassware -- Bottles -- Part 1: Screw-neck bottles (does not apply to 10 ml bottle)
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- Retrace Code, and certificate for manufacturing lot traceability
- Fired-on, highly durable white print
- Suitable for long-term storage
- Easy-to-read scale with graduation marks to estimate the content
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service