Skip to Content
Merck

Skip To

DWK261322108

DURAN® Straight Rim Culture Tube

new

glass tube diam. × H 16 mm × 160 mm, tube capacity 20 mL, borosilicate glass 3.3 tube, neck joint: straight, for Kapsenberg caps, without cap

Synonym(s):

261322108, DURAN tube, cell culture tube, culture tube, tube

Sign Into View Organizational & Contract Pricing

Select a Size

Select a Size

Change View

About This Item

UNSPSC Code:
41121706
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

borosilicate glass 3.3 tube

Joint

joint: neck straight

feature

autoclavable
without cap
for Kapsenberg caps

packaging

pack of 100 ea

manufacturer/tradename

DWK Life Sciences DURAN® 261322108

glass tube diam. × H

16 mm × 160 mm

tube capacity

20 mL

suitability

suitable for (culture)

Looking for similar products? Visit Product Comparison Guide

General description

The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. The original DURAN® culture tubes are distinguished by their uniform wall thickness distribution, minimal thermal expansion and the resulting high resistance to temperature changes. They are therefore particularly suitable for work at very high temperatures and rapid temperature changes.

Application

In vitro cell culture

Features and Benefits

  • Complies with DIN 38411
  • Manufactured from borosilicate glass 3.3
  • Culture tubes with straight edges can be closed with metal caps or cotton plugs to allow air exchange
  • The straight rim permits the use of Kapsenberg and metal caps

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service