Key Documents
GN781280
Greiner microplate
384 well, conical bottom, translucent
Synonym(s):
384 well microplate, 384 well plate, cell culture plate
About This Item
UNSPSC Code:
41121800
material:
conical bottom
size:
384 wells
material
conical bottom
packaging
pack of 100 ea
manufacturer/tradename
Greiner 781280
availability
available only in China
size
384 wells
suitability
suitable for (cell culture)
General description
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.
Features and Benefits
  • Manufactured under DIN ISO 9001 guidelines
  • Can be tracked back to manufacturing using a LOT numbering system.
  • In comparison to the 96 well standard microplate, the number of wells in this microplate is quadrupled, with a volume reduction from 382 ml tο 138 ml
  • 4.5 mm is the well-to-well spacing
  • External dimensions are suitable for use with standard equipment and automated systems.
  • Biomolecular binding characteristics are low
  • Resistant to a wide range of common laboratory chemicals, including DMSO.
  • Has a solid V-bottom (conical bottom)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)