R1527
Monoclonal Anti-Rit1 antibody produced in mouse
~1 mg/mL, clone 14G7, purified immunoglobulin, buffered aqueous glycerol solution
Synonym(s):
Anti-Ras-like protein in tissues
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
14G7, monoclonal
form
buffered aqueous glycerol solution
mol wt
~36 kDa
species reactivity
mouse, human
concentration
~1 mg/mL
technique(s)
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... RIT1(6016)
Immunogen
Target description
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service