Skip to Content
Merck
All Photos(1)

Key Documents

SBR00063

Sigma-Aldrich

Danofloxacin Mesylate

≥98% (HPLC), powder

Synonym(s):

CP-76136-27, Danofloxacin methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24FN3O6S
CAS Number:
Molecular Weight:
453.48
UNSPSC Code:
51101500

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to yellow

mp

>300 °C

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma

Mode of action

DNA synthesis | inhibits

storage temp.

2-8°C

SMILES string

CN1C[C@@H]2C[C@H]1CN2C3=C(C=C4C(=C3)N(C=C(C4=O)C(=O)O)C5CC5)F.CS(=O)(=O)O

InChI

1S/C19H20FN3O3.CH4O3S/c1-21-7-12-4-11(21)8-22(12)17-6-16-13(5-15(17)20)18(24)14(19(25)26)9-23(16)10-2-3-10;1-5(2,3)4/h5-6,9-12H,2-4,7-8H2,1H3,(H,25,26);1H3,(H,2,3,4)/t11-,12-;/m0./s1

InChI key

APFDJSVKQNSTKF-FXMYHANSSA-N

General description

Danofloxacin Mesylate is a synthetic, broad-spectrum fluoroquinolone with antibacterial activity. Danofloxacin Mesylate exerts its antibacterial activity by inhibiting DNA gyrase and topoisomerase IV, enzymes that are essential for bacterial DNA synthesis and replication.

Application

Have been studied for their applications as scaffolds for the development of new antibiotics and other drugs targeting bacterial DNA gyrase and topoisomerase IV.

Biochem/physiol Actions

Danofloxacin Mesylate inhibits bacterial DNA synthesis by interfering with the activity of DNA gyrase and topoisomerase IV enzymes, which are necessary for the replication, transcription, and repair of bacterial DNA. Danofloxacin Mesylate is active against Gram-negative, Gram-positive bacteria, mycoplasma.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service