Skip to Content
Merck
All Photos(1)

Key Documents

SBR00064

Sigma-Aldrich

Ipconazole

≥95% (GC), powder

Synonym(s):

Ipconazole, Ipconazole Isomer 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24ClN3O
CAS Number:
Molecular Weight:
333.86
UNSPSC Code:
51101500

Quality Level

Assay

≥95% (GC)

form

powder

color

white to off-white

antibiotic activity spectrum

fungi

Mode of action

cell membrane | inhibits

storage temp.

2-8°C

SMILES string

CC(C)C1CCC(C1(CN2C=NC=N2)O)CC3=CC=C(C=C3)Cl

InChI

InChI=1S/C18H24ClN3O/c1-13(2)17-8-5-15(9-14-3-6-16(19)7-4-14)18(17,23)10-22-12-20-11-21-22/h3-4,6-7,11-13,15,17,23H,5,8-10H2,1-2H3

InChI key

QTYCMDBMOLSEAM-UHFFFAOYSA-N

General description

Ipconazole is an antimicrobial agent that destroys fungi and belongs to a class of compounds called azoles, which inhibit the synthesis of ergosterol, a key component of fungal cell membranes. By disrupting the structure and function of fungal cell membranes, Ipconazole is able to stop the growth and reproduction of fungi.

Application

Pconazole has been investigated for its potential use in the synthesis of new antifungal compounds, as well as for its antifungal mechanism of action and structure-activity relationships.

Biochem/physiol Actions

Ipconazole inhibits the synthesis of ergosterol, a component of fungal cell membranes, leading to the formation of defective fungal cell membranes and causing fungal cell death. Ipconazole is active against Fungi, including including Aspergillus, Blastomyces, Candida, Coccidioides, Histoplasma, and Sporothrix species.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Target Organs

Eyes,Skin,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service