Skip to Content
Merck
All Photos(1)

Key Documents

SML4081

Lotilaner

≥98% (HPLC)

Synonym(s):

5-[(5S)-4,5-Dihydro-5-(3,4,5-trichlorophenyl)-5-(trifluoromethyl)-3-isoxazolyl]-3-methyl-N-[2-oxo-2-[(2,2,2-trifluoroethyl)amino]ethyl]-2-thiophenecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14Cl3F6N3O3S
CAS Number:
Molecular Weight:
596.76
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

FC(F)(F)[C@@]2(ONC(=C2)c3[s]c(c(c3)C)C(=O)NCC(=O)NCC(F)(F)F)c1cc(c(c(c1)Cl)Cl)Cl

InChI

1S/C20H14Cl3F6N3O3S/c1-8-2-13(36-16(8)17(34)30-6-14(33)31-7-19(24,25)26)12-5-18(35-32-12,20(27,28)29)9-3-10(21)15(23)11(22)4-9/h2-5,32H,6-7H2,1H3,(H,30,34)(H,31,33)/t18-/m0/s1

InChI key

DPFOXZCPAGCTER-SFHVURJKSA-N

Biochem/physiol Actions

Orally available and potent inhibitor of γ-aminobutyric acid (GABA)-gated chloride channels; parasiticide.
Lotilaner is an oral isooxazoline ectoparasitic veterinary medication used to control fleas and ticks in dogs and cats. Lotilaner, similarly to other isoxazolines, targets gamma-aminobutyric acid (GABA) receptors. It binds to the GABA-gated chloride channels in nerve and muscle cells, which blocks the transmission of neuronal signals and leads to paralysis and death.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service