Skip to Content
Merck

SRP0725

Sigma-Aldrich

MLK4, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

15-25 nmol/min·mg (radiometric)

mol wt

~61 kDa

concentration

0.1 μg/μL

accession no.

NM_032435

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MLK4(84451)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant human MLK4 (114-420) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. MLK4 or mixed lineage kinase 4 belongs to the superfamily of MAP kinase kinase kinases (MAP3K1) which contain both ser/thr and tyr kinases activity in their catalytic domains. The structure of this kinase family incorporates an N-terminal Src homology (SH3) domain, followed by the kinase domain, a leucine zipper region, and a CDC42 /RAC -interactive binding (CRIB) motif, but divergent C-terminal regions. MLK4 is highly expressed in kidney and pancreas (1). MLK4 is a negative regulator of TLR4 (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service