Skip to Content
Merck
All Photos(1)

Key Documents

Z332127

BRAND® Dispensette® III bottle-top dispenser, fixed volume

volume 5 mL, without SafetyPrime

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

feature

autoclavable
DE-M marking
without SafetyPrime

manufacturer/tradename

BRAND 4700230 EU
BrandTech 4701230 US

volume measuring range

accuracy: 0.5%, precision: 0.1%

volume

5 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Reliable and long lasting through high-grade materials such as ETFE, PTFE, PFA, FEP, Borosilicate glass and Platinum-Iridium
  • Safety discharge system. Minimises inadvertent dispensing if discharge tube is improperly installed
  • Adjustable dispensing angle. The discharge tube can be positioned in any direction so that the bottle label is always visible. Discharge safety cap
  • Telescopic filling tube. Adjusts easily to different size bottles - without measuring and cutting
  • Maintenance friendly ball valve

Dispensette III: Ideal for dispening aggressive reagents including strong acids e.g. H3PO4,H2SO4, bases like NaOH, KOH, saline solutions and many solvents (e.g. methanol, acetonitrile).

If fitted with SafetyPrime recirculation valve, avoids reagent waste by recirculating during priming.

Features and Benefits

  • Simple: Wide range of applications for organic solvents, acids, alkalis and saline solutions.
  • Easy: Minimum operating forces, maximum comfort.
  • Efficient: Reliable in challenging continuous operation and with aggressive media.

Linkage

See the Compatibility Chart to choose the correct bottle-top dispenser.

Other Notes

BR4700230 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service