Z744245
Wilmad® 5 mm Finned Vortex Plug
PTFE, pack of 1 ea
Synonym(s):
NMR positioning plug, NMR sample processing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.15
Recommended Products
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-805
General description
Plugs are made of PTFE and chemically resistent to most orgianic compound. Recommended for sample positioning inside the NMR tube.
Application
NMR
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service