797-0225
GC Analysis of 4-Methyloctanoic Acid Enantiomers on γ-DEX™ 120
suitable for GC
technique(s)
gas chromatography (GC): suitable
test parameters
column: γ-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24307)
oven: 115 °C
inj. temp.: 220 °C
detector: FID, 300 °C
carrier gas: helium, 20 cm/sec
injection: 1 μL, 100:1 split
sample: ~1 mg/mL each enantiomer in methylene chloride
application(s)
chemicals and industrial polymers
food and beverages
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service