Skip to Content
Merck
Corning® Axygen® 16 Well PCR Microplate 16 well

Corning® Axygen® 16 Well PCR Microplate

Corning® Axygen® 16 Well PCR Microplate 16 well
Synonym(s):
Corning® Axygen® PCR Microplate

Products

Product Category
Manufacturer
Wells
Working Volume (ml)
Color
Package Size
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing