AXYPCR16C
Corning® Axygen® 16 Well PCR Microplate
16 well
Synonym(s):
Corning® Axygen® PCR Microplate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear plate
clear polypropylene wells
clear polypropylene wells
size:
96 wells
Recommended Products
material
clear plate
clear polypropylene wells
sterility
non-sterile
packaging
case of 50 ea
manufacturer/tradename
Axygen Inc
Corning
technique(s)
PCR: suitable
size
96 wells
working volume
200 mL
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service