Skip to Content
Merck

MABS2439

Anti-Adiponectin Antibody, clone AD72

new

Synonym(s):

30 kDa adipocyte complement-related protein, Adipocyte complement-related 30 kDa protein (ACRP30), Adipocyte, C1q and collagen domain-containing protein, Adipose most abundant gene transcript 1 protein, apM-1, Gelatin-binding protein, ACDC, ACRP30, APM1,

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AD72, monoclonal

mol wt

calculated mol wt 26.41 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... ADIPOQ(9370)

Specificity

Clone AD72 is a mouse recombinant monoclonal antibody that detects Adiponectin. Suitable for ELISA application paired with Anti-Adiponectin, clone AD105 (MABS2438).

Immunogen

hFc-tagged, full length, human recombinant Adiponectin.

Application

Quality Control Testing

Evaluated by ELISA with hFc-tagged, full length, human recombinant Adiponectin.


Enzyme Immunoassay (ELISA): Various dilutions from a representative lot of this antibody detected hFc- tagged, recombinant human Adiponectin in ELISA application.

Target description

Adiponectin (UniProt: Q15848; also known as 30 kDa adipocyte complement-related protein, Adipocyte complement-related 30 kDa protein (ACRP30), Adipocyte, C1q and collagen domain-containing protein, Adipose most abundant gene transcript 1 protein, apM-1, Gelatin-binding protein) is encoded by the ADIPOQ (also known as ACDC, ACRP30, APM1, GBP28) gene (Gene ID: 9370) in human. Adiponectin is a homomultimeric secreted glycoprotein, characterized by a collagen-like domain (aa 42-107) and a globular, C1q domain (aa 108-244). It exists in several oligomeric forms, including trimers (low-molecular-weight (LMW)), hexamers, and 12 to 18-mers (high-molecular-weight (HMW)) complexes, with the HMW form being the most biologically active. Adiponectin enhances insulin sensitivity, regulate glucose metabolism, and promote fatty acid oxidation. It activates various signaling pathways, notably the AMP-activated protein kinase (AMPK) pathway, leading to improved metabolic health. Additionally, adiponectin has anti-inflammatory effects and plays a role in cardiovascular health. Adiponectin is expressed in adipose tissue and secreted into the bloodstream. Low levels of adiponectin are linked to obesity, type 2 diabetes, and cardiovascular diseases. Its anti-inflammatory properties suggest a protective role against atherosclerosis and other inflammatory conditions.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library