Skip to Content
Merck

SAB1309054

[KO Validated]Anti-Presenilin 1 antibody produced in rabbit

new

[Knockout Validated],purified polyclonal antibody, buffered aqueous solution

Synonym(s):

Presenilin-1, PS-1, 3423-, Protein S182, Presenilin-1 NTF subunit, Presenilin-1 CTF subunit, Presenilin-1 CTF12, PS1-CTF12, PSEN1, AD3, PS1, PSNL1

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

(Human)

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

280739Da

species reactivity

mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50~100
western blot: 1:1000
western blot: 1:500-1:2000

isotype

IgG

NCBI accession no.

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... PSEN1(5663)

General description

Alzheimer′s disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1; PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor, such that they either directly regulate gamma-secretase activity or themselves are protease enzymes.

Specificity

This Presenilin 1 (PSEN1) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 330-359 amino acids from the C-terminal region of human Presenilin 1 (PSEN1).

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service