SAB5702478
Anti-PAK2 Antibody, clone 2Y7I2, Rabbit Monoclonal
Synonym(s):
PAK65, PAKgamma
biological source
rabbit
Quality Level
material
colorless
clone
2Y7I2, monoclonal
form
liquid
mol wt
61 kDa
species reactivity
rat, rat, human
concentration
0.53 mg/mL
technique(s)
immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000
color
colorless
isotype
IgG
immunogen sequence
MSDNGELEDKPPAPPVRMSSTIFSTGGKDPLSANHSLKPLPSVPEEKKPRHKIISIFSGTEKGSKKKEKERPEISPPSDFEHTIHVGFDAVTGEFTGMPE
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PAK2(5062)
General description
The p21 activated kinases (PAK) are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. The PAK proteins are a family of serine/threonine kinases that serve as targets for the small GTP binding proteins, CDC42 and RAC1, and have been implicated in a wide range of biological activities. The protein encoded by this gene is activated by proteolytic cleavage during caspase-mediated apoptosis, and may play a role in regulating the apoptotic events in the dying cell. [provided by RefSeq, Jul 2008]
Immunogen
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PAK2 (Q13177).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Application
WB, IF/ICC
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.