716189
Phenyl vinyl-1,2-13C sulfone
99 atom % 13C
Synonym(s):
Phenyl vinyl-1,2-13C2 sulfone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5SO213CH=13CH2
CAS Number:
Molecular Weight:
170.20
MDL number:
UNSPSC Code:
12352005
Recommended Products
isotopic purity
99 atom % 13C
form
solid
mp
67-69 °C
mass shift
M+2
SMILES string
[13CH2]=[13CH]S(=O)(=O)c1ccccc1
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Legal Information
Licensed from Highlands Stable Isotopes Corp.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service