Skip to Content
Merck
All Photos(1)

Key Documents

939587

Sigma-Aldrich

Lithium nickel manganese cobalt oxide

new

lithium-niobate coated NMC622 cathode active material

Synonym(s):

LiNbO3-coated NMC622, NMC622

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiNi0.6Mn0.2Co0.2O2
CAS Number:
Molecular Weight:
96.93
UNSPSC Code:
26111700

grade

battery grade

Quality Level

description

application(s): battery manufacturing

Assay

99% trace metals basis

form

powder

mol wt

96.93 g/mol

composition

LiNi0.6Mn0.2Co0.2O2

density

4.8 g/cm3 ((lit.))

General description

Lithium niobate coated NMC622 is a powdered form of lithium nickel manganese cobalt oxide with a stoichiometry of 6:2:2 Ni:Mn:Co and a typical particle size of 4-10 microns. The particles are coated in 5-10 nm amorphous lithium niobate, less than 1 wt.%.

Application

LiNbO3-coated NMC622 is a high-performance cathode active material specially designed for lithium-ion batteries (LIBs), particularly solid-state and high-voltage batteries. With a balance of 60% nickel for high capacity and 40% manganese and cobalt for structural stability, NMC622 surpasses NMC811 in terms of cycling lifetime and coulombic efficiencies and offers higher capacity than NMC532. The LiNbO3 coating plays a vital role in enhancing the stability of the cathode material by safeguarding the electrode and electrolyte from undesired reactions. Research suggests that this lithium-niobate coating mitigates reactions between the cathode material and electrolyte, reducing capacity fade over time and impeding the release of metal ions from the NMC cathode. Additionally, the LiNbO3 “insertion layer” improves rate capability by minimizing the space charge layer barrier at the electrolyte/cathode interface. This advancement translates to superior Li diffusion and reduced electrode polarization.

Features and Benefits

•NMC622: High capacity and long cycle lifetimes
•LiNbO3 coating: Enhanced protection for cathode and electrolyte, leading to improved coulombic efficiency
•LiNbO3 insertion layer: Boosts rate capability

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service