Skip to Content
Merck
All Photos(1)

Key Documents

939846

Sigma-Aldrich

NanoFabTx phenyl boronate functionalized PAMAM dendrimer

new

20 ml/ml aqueous solution, for protein delivery

Synonym(s):

Poly(amidoamine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=5); dendri PAMAM(NH2)128
UNSPSC Code:
12352200

Quality Level

form

liquid

concentration

15-25 mg/mL

color

colorless

density

1.005 g/mL

storage temp.

2-8°C

General description

About this material:

The NanoFabTx Phenyl Boronate Functionalized PAMAM Dendrimer is a specialized drug delivery solution developed and tested for efficient protein delivery. This dendrimer exhibits a high binding affinity with a wide range of proteins, thanks to its rich phenylboronic acid (PBA) content. By leveraging the electron-deficient nature of phenyl boronic acid, this dendrimer can effectively coordinate with cationic amine and imidazole groups on proteins, enabling nitrogen-boronate complexations and cationic and ionic interactions. Moreover, the PBA moieties on this NanoFabTx dendrimer provide opportunities for self-assembly and effectively address existing barriers in the cytosolic delivery of nucleic acids.

More information including application notes can be found under the "More Documents" tab on this page.

Application

Specifically optimized for intracellular protein delivery, including protein complexes, the NanoFabTx(TM) phenyl boronate functionalized PAMAM dendrimer has applications in:

  • Therapuetic nuclelic acid and protein delivery - This dendrimer enables the formation of dendrimer-protein complexes thatcan bypass membrane barriers, enable high uptake by mammalian cells, and aid in cysoloic delivery into the cells
  • Molecular and cell detection: The high density of boronic groups can be used fro molecular and cell detections such as sugar and H2O2

Features and Benefits

The NanoFabTx drug delivery solutions have been tested to ensure effective therapeutic delivery, and they are accompanied by supported protocols and application notes to guide you on your drug delivery journey. Derived from Sigma-Aldrich Product 536709 (ethylenediamine core, generation 5.0), the NanoFabTx phenyl boronate functionalized PAMAM dendrimer offers the following features and benefits:

  • A ready-to-use formulation for synthesizing dendrimer-protein complexes
  • Detailed, step-by-step protocols for protein delivery, along with application data for in vitro delivery
  • A drug delivery solution that enables protein delivery into cells without noticeable toxicity or disruption of metabolism
  • Flexibility to optimize the material based on protein size and surface properties, providing adaptability for various applications and research needs.
For research use only NanoFabTx is a trademark of Sigma-Aldrich Co. LLC

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service